MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Long-term follow-up: no effect of therapeutic vaccination with HIV-1 p17/p24:Ty virus-like particles on HIV-1 disease progression.
Long-term follow-up: no effect of therapeutic vaccination with HIV-1 p17/p24:Ty virus-like particles on HIV-1 disease progression.
Vaccine (2002-05-16)
Catharina E A Lindenburg, Ineke Stolte, Miranda W Langendam, Frank Miedema, Ian G Williams, Robert Colebunders, Jonathan N Weber, Martin Fisher, Roel A Coutinho
RESUMEN
Long-term effects of therapeutic vaccination of human immunodeficiency virus (HIV)-1-infected subjects with HIV-1 p17/p24:Ty virus-like particles (p24-VLP) on progression to AIDS, death, a CD4 cell count <or=200 cells/mm(3) and CD4 cell count decline were studied in a multicenter cohort study of 56 individuals who participated in a phase II double-blind placebo-controlled trial with p24-VLP in 1993. Using Cox proportional hazard analysis, no difference between vaccine and placebo groups was found in progression to death (adjusted hazard rate (HR): 0.68 (95% CI: 0.05-7.83), AIDS (adjusted HR: 1.07 (95% CI: 0.21-5.36)) and a CD4 cell count <or=200 cells/mm(3) (adjusted HR: 1.00 (95% CI: 0.35-2.87)). Using linear regression with correction for multiple visits within one person, no effect of vaccination on CD4 cell count decline, adjusted for antiretroviral therapy (ART) use, was found (P=0.98). In conclusion, therapeutic vaccination with p24-VLP is not related to slower HIV-1 disease progression.