MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Preparation, characterization, and evaluation in vivo of Ins-SiO₂-HP55 (insulin-loaded silica coating HP55) for oral delivery of insulin.
Preparation, characterization, and evaluation in vivo of Ins-SiO₂-HP55 (insulin-loaded silica coating HP55) for oral delivery of insulin.
International journal of pharmaceutics (2013-07-09)
Xiuhua Zhao, Chang Shan, Yuangang Zu, Ying Zhang, Weiguo Wang, Kunlun Wang, Xiaoyu Sui, Ruiqiang Li
RESUMEN
Insulin is the most effective and durable drug in the treatment of advanced stage diabetes. However, oral delivering insulin was a tough task for rapid enzymatic degradation. In this work, we designed and developed a delivery system composed of enteric nanosphere for oral delivery of insulin. The silica was selected for loading insulin, which surface has a lot of pores with a powerful adsorption capacity, advantages for permeability and slow-release. The insulin-loaded silica (Ins-SiO2) was prepared by adsorption in HCl solution. The Ins-SiO2 obtained was coated with the hydroxypropyl methylcellulose phthalate (HP55) by desolvation method, which is a good enteric coating material. The Ins-SiO2-HP55, an enteric nanosphere of insulin obtained were characterized by transmission electron microscope (TEM), surface area, Fourier-transform infrared (FT-IR), X-ray diffraction (XRD), differential scanning calorimetry (DSC) and thermogravimetric analysis (TGA). The results showed that insulin was loaded most in the pores of silica, while the HP55 coated on the extent of Ins-SiO2. In vitro drug release results revealed that the release of insulin from Ins-SiO2-HP55 was markedly reduced in simulated gastric fluid (SGF). By contrast, the release amount of insulin from Ins-SiO2-HP55 was increased significantly in simulated intestinal fluid (SIF). In vivo evaluation on diabetic animals showed the blood glucose level of diabetic rats could be effectively reduced after oral administration Ins-SiO2-HP55. There is marked hypoglycemic effect after 1h of taking the Ins-SiO2-HP55. After 3h, the GLU of rats of the Ins-SiO2-HP55 stably kept from 4.85 to 2.67 mmol/L that was significantly less than the normal level (6.7 mmol/L). However, that of rats taking raw insulin kept from 8.03 to 6.56 mmol/L that is higher than the normal level. These results suggested that Ins-SiO2-HP55 could have potential value in oral administration systems of diabetes chemotherapy.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP