MilliporeSigma
  • Effects of switching from topical beta-blockers to latanoprost on intraocular pressure in patients with normal-tension glaucoma.
Effects of switching from topical beta-blockers to latanoprost on intraocular pressure in patients with normal-tension glaucoma.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics (2008-03-18)
Yoko Ikeda, Kazuhiko Mori, Takeshi Ishibashi, Shigeta Naruse, Nobuko Nakajima, Shigeru Kinoshita
RESUMEN
The effects of switching from topical beta-blockers (beta) to latanoprost (LA) on intraocular pressure (IOP) and IOP-reduction rate (IOP-RR) in patients with normal-tension glaucoma (NTG) were investigated. Sixty (60) NTG patients (60 eyes) were divided into three equal groups receiving carteolol hydrochloride (group A), nipradilol (group B), and betaxolol hydrochloride (group C) twice-daily for 3 months. The drugs were changed to topical LA administered once-daily for the next 3 months. Baseline IOP was 14.4 +/- 0.9, 14.6 +/- 0.6, and 14.6 +/- 0.9 mmHg in groups A, B, and C, respectively. At 3 months, IOP was 12.4 +/- 0.6, 13.4 +/- 0.6, and 12.9 +/- 0.8 mmHg and 10.5 +/- 0.5, 11.1 +/- 0.8, and 11.7 +/- 0.8 mmHg at 6 months in groups A, B, and C, respectively. At 3 months, IOP-RR was 10.4 +/- 5.5, 9.5 +/- 2.6, and 10.8 +/- 4.7% and 24.1 +/- 4.3, 22.9 +/- 5.9, and 19.4 +/- 3.8% at 6 months in groups A, B, and C, respectively. The groups did not significantly differ in the first 3 months regarding IOP and IOP-RR. Switching to LA significantly decreased IOP and increased IOP-RR in all groups. In NTG patients, LA reduced IOP more effectively than the beta tested.
MATERIALES
Número de producto
Marca
Descripción del producto
Carteolol for system suitability, European Pharmacopoeia (EP) Reference Standard