MilliporeSigma
Co-processed MCC-Eudragit® E excipients for extrusion-spheronization.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2011-09-29)
Alvaro Goyanes, Consuelo Souto, Ramón Martínez-Pacheco
RESUMEN
This study investigates the extrusion-spheronization performance of some mixtures of co-processed microcrystalline cellulose and Eudragit® E (as excipients) and sorbitol (as soluble filler-disintegrant). Attention is focused on the dissolution rate of low water solubility drugs (hydrochlorothiazide is used as a model drug) from pellets prepared with these mixtures. All pellet formulations studied presented adequate morphological, flow and mechanical properties. The pellets prepared with co-processed MCC-Eudragit® E and sorbitol show a drug dissolution rate dependent on the content of Eudragit® E in the co-processed excipient and on the proportion of sorbitol incorporated. Furthermore, the pellets made with co-processed MCC-Eudragit® E incorporating the higher proportion of sorbitol (50%) show a very high dissolution rate of hydrochlorothiazide (HCT) and undergo rapid disintegration in the dissolution medium.
MATERIALES
Número de producto
Marca
Descripción del producto
Basic butylated methacrylate copolyme, European Pharmacopoeia (EP) Reference Standard