MilliporeSigma
Effect of the phytotherapeutic agent eviprostat on the bladder in a rat model of bladder overdistension/emptying.
Neurourology and urodynamics (2012-11-13)
Yuko Kawai, Michiko Oka, Junko Kyotani, Tatsuya Oyama, Seiji Matsumoto, Hidehiro Kakizaki
RESUMEN
Ischemia-reperfusion injury is an important factor in the development of lower urinary tract symptoms (LUTS) that is partly mediated by the generation of free radicals. We investigate the effect of the phytotherapeutic agent Eviprostat, a treatment for benign prostatic hyperplasia (BPH) that has antioxidant and anti-inflammatory activity, on urinary bladder blood flow (BBF), and function in a rat model of bladder overdistension and emptying (OE). For 8 days before surgery, OE rats received daily oral Eviprostat (36 mg/kg/day) or vehicle, while sham-operated animals received vehicle. The bladder was distended by infusion of saline over a period of 2 hr (overdistension) and then emptied. After 24 hr, BBF was measured with a laser speckle blood flow imager. The oxidative-stress marker malondialdehyde (MDA), proinflammatory cytokines, and myeloperoxidase were determined in the isolated bladder, and histological analysis was performed. Functional contractile responses of bladder strips to electrical field stimulation, carbachol, and KCl were measured. Twenty-four hours after bladder OE, a significant decrease in BBF and significant increases in bladder weight, malondialdehyde, proinflammatory cytokines, and myeloperoxidase were observed. Eviprostat almost completely prevented these changes. Histological analysis of the bladder of OE rats showed hemorrhage, accumulation of leukocytes, desquamation of epithelium, and edema, and Eviprostat suppressed these changes. The reduction in functional contractile forces in the bladder of OE rats was also prevented by Eviprostat. Eviprostat-mediated suppression of increased bladder oxidative stress and inflammation caused by bladder OE may contribute to the improvement of BBF and bladder function by Eviprostat.
MATERIALES
Número de producto
Marca
Descripción del producto
Etamsylate, European Pharmacopoeia (EP) Reference Standard