MilliporeSigma
  • A sensitive assay of metoprolol and its major metabolite alpha-hydroxy metoprolol in human plasma and determination of dextromethorphan and its metabolite dextrorphan in urine with high performance liquid chromatography and fluorometric detection.
A sensitive assay of metoprolol and its major metabolite alpha-hydroxy metoprolol in human plasma and determination of dextromethorphan and its metabolite dextrorphan in urine with high performance liquid chromatography and fluorometric detection.
Journal of pharmaceutical and biomedical analysis (1998-04-21)
B Mistry, J Leslie, N E Eddington
RESUMEN
A reverse-phase High Performance Liquid Chromatographic (HPLC) method was developed for the analysis of metoprolol in the large number of human plasma samples obtained in in vitro-in vivo correlations (IVIVC) and bioavailability studies of extended release formulations of metoprolol tartrate. The metabolite, alpha-hydroxy metoprolol (OH-met), could also be quantified. The analytes were extracted from the plasma using solid phase columns, separated on a C-4 analytical column followed by fluorimetric detection. The linearity, precision, accuracy, stability, selectivity and ruggedness were validated for the concentration ranges of 1-400 ng ml-1 for metoprolol and 0.5-200 ng ml-1 for OH-met. The same chromatographic conditions were slightly modified to quantify dextromethorphan and its metabolite dextrorphan in urine in the concentration range 0.052-0.05 microgram ml-1 as a method for screening for fast metabolizers.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
α-Hydroxymetoprolol, analytical standard