MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • European recommendations for primary prevention of congenital anomalies: a joined effort of EUROCAT and EUROPLAN projects to facilitate inclusion of this topic in the National Rare Disease Plans.
European recommendations for primary prevention of congenital anomalies: a joined effort of EUROCAT and EUROPLAN projects to facilitate inclusion of this topic in the National Rare Disease Plans.
Public health genomics (2014-04-10)
Domenica Taruscio, Larraitz Arriola, Francesca Baldi, Ingeborg Barisic, Eva Bermejo-Sánchez, Fabrizio Bianchi, Elisa Calzolari, Pietro Carbone, Rhonda Curran, Ester Garne, Miriam Gatt, Anna Latos-Bieleńska, Babak Khoshnood, Lorentz Irgens, Alberto Mantovani, Maria Luisa Martínez-Frías, Amanda Neville, Anke Rißmann, Stefania Ruggeri, Diana Wellesley, Helen Dolk
RESUMEN
Congenital anomalies (CA) are the paradigm example of rare diseases liable to primary prevention actions due to the multifactorial etiology of many of them, involving a number of environmental factors together with genetic predispositions. Yet despite the preventive potential, lack of attention to an integrated preventive strategy has led to the prevalence of CA remaining relatively stable in recent decades. The 2 European projects, EUROCAT and EUROPLAN, have joined efforts to provide the first science-based and comprehensive set of recommendations for the primary prevention of CA in the European Union. The resulting EUROCAT-EUROPLAN 'Recommendations on Policies to Be Considered for the Primary Prevention of Congenital Anomalies in National Plans and Strategies on Rare Diseases' were issued in 2012 and endorsed by EUCERD (European Union Committee of Experts on Rare Diseases) in 2013. The recommendations exploit interdisciplinary expertise encompassing drugs, diet, lifestyles, maternal health status, and the environment. The recommendations include evidence-based actions aimed at reducing risk factors and at increasing protective factors and behaviors at both individual and population level. Moreover, consideration is given to topics specifically related to CA (e.g. folate status, teratogens) as well as of broad public health impact (e.g. obesity, smoking) which call for specific attention to their relevance in the pre- and periconceptional period. The recommendations, reported entirely in this paper, are a comprehensive tool to implement primary prevention into national policies on rare diseases in Europe.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Supelco
Folic acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Folic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Folic acid, meets USP testing specifications
Folic acid, European Pharmacopoeia (EP) Reference Standard