MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Prenatal management with digoxin and sotalol combination for fetal supraventricular tachycardia: case report and review of literature.
Prenatal management with digoxin and sotalol combination for fetal supraventricular tachycardia: case report and review of literature.
Indian journal of medical sciences (2009-10-07)
Vanita Suri, Anish Keepanaseril, Neelam Aggarwal, Rajesh Vijayvergiya
RESUMEN
Sustained fetal supraventricular tachycardia (SVT) with a heart rate of approximately 210 bpm may lead to increased atrial and venous pressures and congestive heart failure. There is no clear consensus regarding the best drug-treatment regimens for fetal SVT. However, considerable nonrandomized experience in the transmaternal treatment of fetal SVT is available with a number of antiarrhythmic agents. We report a case of fetal supraventricular tachyarrhythmia with hydrops detected at 32 weeks that was managed with combination of oral digoxin and sotalol and review management guidelines available in the literature.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Digoxin, analytical standard
USP
Digoxin, United States Pharmacopeia (USP) Reference Standard
Supelco
Digoxin solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Digoxin, European Pharmacopoeia (EP) Reference Standard
Digoxin for peak identification, European Pharmacopoeia (EP) Reference Standard