- Cobalt-catalyzed asymmetric 1,6-addition of (triisopropylsilyl)-acetylene to α,β,γ,δ-unsaturated carbonyl compounds.
Cobalt-catalyzed asymmetric 1,6-addition of (triisopropylsilyl)-acetylene to α,β,γ,δ-unsaturated carbonyl compounds.
Journal of the American Chemical Society (2012-11-08)
Takahiro Sawano, Akram Ashouri, Takahiro Nishimura, Tamio Hayashi
PMID23131028
RESUMEN
Asymmetric addition of (triisopropylsilyl)acetylene to α,β,γ,δ-unsaturated carbonyl compounds took place in the presence of a cobalt/Duphos catalyst to give the 1,6-addition products in high yields with high regio- and enantioselectivity.
MATERIALES