MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Influence of short-term exposure to low levels of 17α-ethynylestradiol on expression of genes involved in immunity and on immune parameters in rainbow trout, Oncorhynchus mykiss.
Influence of short-term exposure to low levels of 17α-ethynylestradiol on expression of genes involved in immunity and on immune parameters in rainbow trout, Oncorhynchus mykiss.
Aquatic toxicology (Amsterdam, Netherlands) (2014-12-03)
Sophie Massart, Sylvain Milla, Baptiste Redivo, Enora Flamion, S N M Mandiki, Elodie Falisse, Patrick Kestemont
RESUMEN
Fish are exposed to endocrine-disrupting chemicals (EDC), which are well known to disturb not only the reproductive system but also the immune system in vertebrates. However, the mechanisms by which these compounds are able to modify fish immunity are not well understood. In order to test the EE2 effects on immunity in selected organs, we exposed rainbow trout male juveniles for 3 weeks to EE2 concentrations ranging from 0.01 to 1 μg/L. The results of this study suggest that EE2 affects the immunity of rainbow trout in a tissue dependent manner. This molecule affects both cellular and humoral immune systems. Indeed, blood leukocyte populations, as well as hepatic and plasma lysozyme, plasma MPO and renal complement activities, are modulated by EE2. Moreover, EE2 alters the gene expression of some mucus compounds, hepatic expression of complement sub-unit and lysozyme, or genes involved in the hepatic phagocytosis and transport of immunoglobulin across the liver.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
17α-etinilestradiol, ≥98%
Supelco
17α-etinilestradiol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Etinil estradiol, United States Pharmacopeia (USP) Reference Standard
17α-etinilestradiol, European Pharmacopoeia (EP) Reference Standard
Supelco
17α-Ethynylestradiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
17α-etinilestradiol, European Pharmacopoeia (EP) Reference Standard