MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Infection of human amniotic and endothelial cells by Japanese encephalitis virus: Increased expression of HLA-F.
Virology (2014-12-03)
Gaurav Kumar, Onkar Sanjay Date, Kwang Sik Kim, Ramanathapuram Manjunath
RESUMEN
Productive infection of human amniotic and endothelial cell lines with Japanese encephalitis virus (JEV) was established leading to the induction of NFκB and HLA-F, a non-classical MHC molecule. Induction of the HLA-F gene and protein in JEV-infected cells was shown to be NFκB dependent since it was blocked by inhibitors of NFκB activation. ShRNA targeting lentivirus-mediated stable knockdown of the p65 subunit of NFκB inhibited JEV-mediated induction of HLA-F both in the amniotic cell line, AV-3 as well as the human brain microendothelial cell line, HBMEC. The induction of HLA-F by treatment of AV-3 with TNF-α was also inhibited by ShRNA mediated knockdown of NFκB. TNF-α treatment of HEK293T cells that were transfected with reporter plasmids under the control of HLA-F enhancer A elements resulted in significant transactivation of the luciferase reporter gene. NFκB-mediated induction of HLA-F following JEV infection and TNF-α exposure is being suggested for the first time.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Glicerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glicerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glicerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Fluoruro de fenilmetansulfonilo, ≥98.5% (GC)
Sigma-Aldrich
Glicerol, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Piruvato sódico, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Glicerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Piruvato sódico, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glicerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Glicerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glicerol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glicerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Fluoruro de fenilmetansulfonilo, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
USP
Glicerol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glicerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glicerol, FCC, FG
Sigma-Aldrich
Glicerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glicerol, meets USP testing specifications
Sigma-Aldrich
Piruvato sódico, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Glicerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Piruvato sódico, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Dibenzothiophene, 98%
USP
Levotiroxina, United States Pharmacopeia (USP) Reference Standard