MilliporeSigma
  • High-speed monitoring of the crystallinity change in poly(lactic acid) during photodegradation by using a newly developed wide area NIR imaging system (Compovision).
High-speed monitoring of the crystallinity change in poly(lactic acid) during photodegradation by using a newly developed wide area NIR imaging system (Compovision).
Analytical and bioanalytical chemistry (2014-10-20)
Daitaro Ishikawa, Daiki Furukawa, Tseng Tsai Wei, Kummetha Raghunatha Reddy, Asako Motomura, Yoko Igarashi, Harumi Sato, Sergei G Kazarian, Yukihiro Ozaki
RESUMEN
We aimed to achieve wide area rapid monitoring of the crystallinity change in poly(lactic acid) (PLA) during photodegradation caused by ultraviolet (UV) light by using a newly developed near-infrared (NIR) camera (Compovison). Several kinds of PLA samples with different crystallinities and their blends with poly[(3)-(R)-hydroxybutyrate] were prepared. Their two-dimensional NIR spectra in the 1,000-2,350-nm region were measured by Compovision at a 5-min interval during photolysis. An intensity decrease of the band in the 1,900-1,925-nm region due to the second overtone of the C = O stretching vibration of PLA was observed during photolysis. This suggests that an anhydride carbonyl is produced during photolysis. The NIR image of the crystallinity change monitored by the band at 1,917 nm in the standard normal variate spectra clearly shows the inhomogeneity of crystal evolution. A logarithmic increase was observed for all identified areas in the PLA film; however, the time to reach the maximum crystallinity was slightly different according to the initial crystallinity of the sample. It is likely that the initial crystallinity of the sample influences the degradation speed more than the degradation amount. These imaging results have provided fundamental chemical insights into the photolytic process for PLA, and at the same time they have demonstrated that the two-dimensional spectral data obtained by Compovision are useful for process monitoring of polymers.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cloroformo, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Cloroformo, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Cloroformo, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Cloroformo, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Cloroformo, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Cloroformo, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Cloroformo, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Cloroformo, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Cloroformo, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Cloroformo, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Cloroformo, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Cloroformo, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Cloroformo, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Cloroformo, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Cloroformo, JIS special grade, ≥99.0%
Supelco
Cloroformo, analytical standard
Supelco
Cloroformo, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cloroformo, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Cloroformo, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cloroformo, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Cloroformo, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Cloroformo, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sigma-Aldrich
Cloroformo, suitable for HPLC
Sigma-Aldrich
Cloroformo, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Cloroformo, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Cloroformo, JIS 300, ≥99.0%