MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Diapocynin, a dimer of the NADPH oxidase inhibitor apocynin, reduces ROS production and prevents force loss in eccentrically contracting dystrophic muscle.
Diapocynin, a dimer of the NADPH oxidase inhibitor apocynin, reduces ROS production and prevents force loss in eccentrically contracting dystrophic muscle.
PloS one (2014-10-21)
Hesham M Ismail, Leonardo Scapozza, Urs T Ruegg, Olivier M Dorchies
RESUMEN
Elevation of intracellular Ca2+, excessive ROS production and increased phospholipase A2 activity contribute to the pathology in dystrophin-deficient muscle. Moreover, Ca2+, ROS and phospholipase A2, in particular iPLA2, are thought to potentiate each other in positive feedback loops. NADPH oxidases (NOX) have been considered as a major source of ROS in muscle and have been reported to be overexpressed in muscles of mdx mice. We report here on our investigations regarding the effect of diapocynin, a dimer of the commonly used NOX inhibitor apocynin, on the activity of iPLA2, Ca2+ handling and ROS generation in dystrophic myotubes. We also examined the effects of diapocynin on force production and recovery ability of isolated EDL muscles exposed to eccentric contractions in vitro, a damaging procedure to which dystrophic muscle is extremely sensitive. In dystrophic myotubes, diapocynin inhibited ROS production, abolished iPLA2 activity and reduced Ca2+ influx through stretch-activated and store-operated channels, two major pathways responsible for excessive Ca2+ entry in dystrophic muscle. Diapocynin also prevented force loss induced by eccentric contractions of mdx muscle close to the value of wild-type muscle and reduced membrane damage as seen by Procion orange dye uptake. These findings support the central role played by NOX-ROS in the pathogenic cascade leading to muscular dystrophy and suggest diapocynin as an effective NOX inhibitor that might be helpful for future therapeutic approaches.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Acetona, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetona, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetona, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Bicarbonato de sodio, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetona, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Bicarbonato de sodio, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Bicarbonato de sodio, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Cloruro de sodio, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de calcio solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Acetona, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetona, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Bicarbonato de sodio, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Bicarbonato de sodio, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Acetona, ACS reagent, ≥99.5%
Supelco
Acetona, analytical standard
Sigma-Aldrich
Cloruro de sodio, JIS special grade, ≥99.5%
Sigma-Aldrich
Cloruro de sodio, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Cloruro de sodio, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O