MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Pharmacokinetic and metabolism studies of rohitukine in rats by high performance liquid-chromatography with tandem mass spectrometry.
Pharmacokinetic and metabolism studies of rohitukine in rats by high performance liquid-chromatography with tandem mass spectrometry.
Fitoterapia (2014-05-21)
Yashpal S Chhonker, Hardik Chandasana, Deepak Kumar, Sunil K Mishra, Shishir Srivastava, Vishal M Balaramnavar, Anil Nilkanth Gaikwad, Sanjeev Kanojiya, Anil K Saxena, Rabi S Bhatta
RESUMEN
A sensitive, selective, and rapid high performance liquid chromatography-tandem mass spectrometry (LC-MS/MS) was developed for the quantification of rohitukine in rat plasma. HPLC was performed using a Symmetry-Shield C18 (5 μ, 4.6 × 150 mm) column, and isocratic elution with ammonium acetate buffer (pH4; 10 mM):methanol (08:92, v/v) at a flow rate of 0.6 mL/min. Sample clean-up involved solid phase extraction (SPE) of analyte and internal standard (phenacetin) from 100 μL plasma. The parent→product ion transitions (MRM) for analyte and IS were 306.1→245.1 m/z and 180.1→138.1 m/z respectively, and were monitored on a triple quadrupole mass spectrometer, operating in positive ion mode. The method was validated across the dynamic concentration range of 5-500 ng/mL for rohitukine, with a fast run time of 4.5 min. The analytical method measured concentrations of rohitukine with accuracy (% bias) of <±10% and precision (% RSD) of <±12%. Rohitukine was stable during the battery of stability studies viz., bench-top, auto-sampler, freeze/thaw cycles and 30 days of storage in a freezer at -70±10°C. Finally, the applicability of this assay has been successfully demonstrated in vivo pharmacokinetic and in vitro metabolism studies in Sprague-Dawley rat. This method will therefore be highly useful for future preclinical and clinical pharmacokinetic studies of rohitukine.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Metanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Metanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Metanol, anhydrous, 99.8%
Sigma-Aldrich
Metanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Metanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Metanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanol, Absolute - Acetone free
USP
Metanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Metanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Metanol, BioReagent, ≥99.93%
Supelco
Metanol, analytical standard
Supelco
Metanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Metanol, SAJ special grade
Supelco
Phenacetin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Metanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
USP
Phenacetin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Metanol, suitable for HPLC
Supelco
Phenacetin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Metanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, 99.93%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Metanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.