MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Low doses of imatinib induce myelopoiesis and enhance host anti-microbial immunity.
PLoS pathogens (2015-03-31)
Ruth J Napier, Brian A Norris, Alyson Swimm, Cynthia R Giver, Wayne A C Harris, Julie Laval, Brooke A Napier, Gopi Patel, Ryan Crump, Zhenghong Peng, William Bornmann, Bali Pulendran, R Mark Buller, David S Weiss, Rabindra Tirouvanziam, Edmund K Waller, Daniel Kalman
RESUMEN
Imatinib mesylate (Gleevec) inhibits Abl1, c-Kit, and related protein tyrosine kinases (PTKs) and serves as a therapeutic for chronic myelogenous leukemia and gastrointestinal stromal tumors. Imatinib also has efficacy against various pathogens, including pathogenic mycobacteria, where it decreases bacterial load in mice, albeit at doses below those used for treating cancer. We report that imatinib at such low doses unexpectedly induces differentiation of hematopoietic stem cells and progenitors in the bone marrow, augments myelopoiesis but not lymphopoiesis, and increases numbers of myeloid cells in blood and spleen. Whereas progenitor differentiation relies on partial inhibition of c-Kit by imatinib, lineage commitment depends upon inhibition of other PTKs. Thus, imatinib mimics "emergency hematopoiesis," a physiological innate immune response to infection. Increasing neutrophil numbers by adoptive transfer sufficed to reduce mycobacterial load, and imatinib reduced bacterial load of Franciscella spp., which do not utilize imatinib-sensitive PTKs for pathogenesis. Thus, potentiation of the immune response by imatinib at low doses may facilitate clearance of diverse microbial pathogens.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-cisteína, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Cloruro de amonio, ReagentPlus®, ≥99.5%
Sigma-Aldrich
L-cisteína, 97%
Sigma-Aldrich
Cloruro de amonio, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
L-cisteína, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Cloruro de amonio, 99.998% trace metals basis
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
SAFC
L-cisteína
Sigma-Aldrich
Iron(III) pyrophosphate, soluble crystals
Supelco
Cloruro de amonio, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-cisteína, ≥97%, FG
Sigma-Aldrich
Cloruro de amonio, 99.99% trace metals basis
Sigma-Aldrich
Cloruro de amonio, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
Supelco
L-cisteína, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Cloruro de amonio, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-cisteína, Wacker Chemie AG, ≥98.0%
Supelco
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Cloruro de amonio, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Supelco
Aphidicolin, analytical standard