MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Cytotoxicity of coated and uncoated fibre-reinforced composites.
Acta odontologica Scandinavica (2014-01-23)
Cornelia Frese, Diana Wolff, Sebastian Zingler, Tanja Krueger, Kathrin Stucke, Christopher J Lux, Hans Joerg Staehle, Ralf Erber
RESUMEN
Currently, there are many fibre-reinforced composites (FRCs) available which differ in the type and volume fraction of fibres, pre-treatment of fibres and matrix composition. The aims of this in vitro investigation were to determine whether there is a difference in biocompatibility of FRCs and if coating FRCs with resin composites influences their cytotoxic potential. Five different FRC materials were tested which were either uncoated or coated with flowable or viscous resin composite. Artificial saliva extracts were prepared according to USP-XXIII and ISO-10993 to determine cytotoxicity by testing cell viability and growth of primary human gingival fibroblasts (HGF) using MTT assay, LIVE/DEAD(®) assay and cell proliferation assay. The influence of eluates on fibres of the cytoskeleton was investigated by vimentin, tubulin and actinin immunostainings. A two-way ANOVA followed by Scheffe's post-hoc test, which included the factors FRC material and coating procedure, was performed to assess cytotoxicity. All extracts of FRC materials displayed minor cytotoxic potential on HGF cell viability, cell proliferation and integrity of the cytoskeleton. The type of FRC material significantly influenced cell viability (MTT assay) (p < 0.0001), whereas neither the presence of a coating nor the type of coating material resulted in altered cell viability. Distribution and organization of cytosolic fibres was not affected after HGF exposure to eluates. There is a lack of knowledge about the leaching behaviour of commonly available fully pre-impregnated FRCs and their interactions with coating materials. The coating of FRCs with resin composite materials did not impact biocompatibility.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cloruro de sodio, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Cloruro de sodio, JIS special grade, ≥99.5%
Sigma-Aldrich
Cloruro de sodio, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Cloruro de sodio, 99.999% trace metals basis
Supelco
Cloruro de sodio, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cloruro de sodio, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Cloruro de sodio, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Cloruro de sodio, SAJ first grade, ≥99.0%
Sigma-Aldrich
Cloruro de sodio, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Cloruro de sodio, tablet
Sigma-Aldrich
Cloruro de sodio, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Cloruro de sodio, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.1 M