MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Influence of micropattern width on differentiation of human mesenchymal stem cells to vascular smooth muscle cells.
Colloids and surfaces. B, Biointerfaces (2014-07-30)
Tomoko Nakamoto, Xinlong Wang, Naoki Kawazoe, Guoping Chen
RESUMEN
In recent years, various approaches have been taken to generate functional muscle tissue by tissue engineering. However, in vitro methods to generate smooth muscle with physiologically aligned structure remains limited. In order to mimic the in vivo highly organized structure of smooth muscle cells, we used micropatterning technology for engineering parallel aligned cells. In this study, a gradient micropattern of different width of cell-adhesive polystyrene stripes (5, 10, 20, 40, 60, 80, 100, 200, 400, 600, 800 and 1000μm) was prepared and the effects of micropattern width on human mesenchymal stem cells (hMSCs) orientation, morphology and smooth muscle cell differentiation were investigated. The width of micropattern stripes showed obvious effect on cell orientation, morphology and smooth muscle cell differentiation. The cells showed higher degree of orientation when the micropattern stripes became narrower. Higher expression of calponin and smooth muscle actin was observed among the narrow micropatterns ranging from 200μm to 20μm, compared to the non-patterned area and wide micropattern areas which showed similar levels of expression.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
DCC, 1.0 M in methylene chloride
Sigma-Aldrich
DCC, puriss., ≥99.0% (GC)