MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Size dependence of ultrafast charge dynamics in monodisperse Au nanoparticles supported on TiO₂ colloidal spheres.
Physical chemistry chemical physics : PCCP (2014-06-10)
Ali Al-Otaify, Marina A Leontiadou, Flavia V E dos Reis, Tatiana C Damato, Pedro H C Camargo, David J Binks
RESUMEN
Sub-nanosecond charge dynamics in monodisperse Au nanoparticles (NPs) supported on TiO2 colloidal spheres are studied as a function of NP diameter using ultrafast transient absorption spectroscopy. The decay of the transmittance changes observed in the region of the plasmon resonance of the Au NPs following photoexcitation of the TiO2 spheres are well-described by a bi-exponential function consisting of a fast component of 2 ps duration associated with electron-phonon scattering, followed by a slow and relatively weak component associated with phonon-phonon scattering. The decay constant characterising the latter component was found to be dependent on the size of the Au NPs, rising from 49 ± 3 to 128 ± 6 ps as the diameter of the Au NPs increased from 12.2 ± 2.2 nm to 24.5 ± 2.8 nm, respectively.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Acetona, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetona, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetona, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetona, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Acetona, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetona, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Butóxido de titanio (IV), reagent grade, 97%
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetona, ACS reagent, ≥99.5%
Supelco
Acetona, analytical standard
Sigma-Aldrich
Acetona, histological grade, ≥99.5%
Sigma-Aldrich
Acetona, JIS special grade, ≥99.5%
USP
Acetona, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethylene glycol, analytical standard
Supelco
Acetona, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetona, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
Acetona, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetona, ≥99%, FCC, FG
Sigma-Aldrich
Butóxido de titanio (IV), purum, ≥97.0% (gravimetric)
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetona, natural, ≥97%
Sigma-Aldrich
Acetona, suitable for HPLC, ≥99.9%
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Acetona, suitable for residue analysis, ≥99.5%
Sigma-Aldrich
Acetona, suitable for HPLC