MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Roles of Cav3.2 and TRPA1 channels targeted by hydrogen sulfide in pancreatic nociceptive processing in mice with or without acute pancreatitis.
Roles of Cav3.2 and TRPA1 channels targeted by hydrogen sulfide in pancreatic nociceptive processing in mice with or without acute pancreatitis.
Journal of neuroscience research (2014-10-01)
Yuka Terada, Mayuko Fujimura, Sachiyo Nishimura, Maho Tsubota, Fumiko Sekiguchi, Atsufumi Kawabata
RESUMEN
Hydrogen sulfide (H(2)S), formed by multiple enzymes, including cystathionine-γ-lyase (CSE), targets Ca(v)3.2 T-type Ca(2+) channels (T channels) and transient receptor potential ankyrin-1 (TRPA1), facilitating somatic pain. Pancreatitis-related pain also appears to involve activation of T channels by H(2)S formed by the upregulated CSE. Therefore, this study investigates the roles of the Ca(v)3.2 isoform and/or TRPA1 in pancreatic nociception in the absence and presence of pancreatitis. In anesthetized mice, AP18, a TRPA1 inhibitor, abolished the Fos expression in the spinal dorsal horn caused by injection of a TRPA1 agonist into the pancreatic duct. As did mibefradil, a T-channel inhibitor, in our previous report, AP18 prevented the Fos expression following ductal NaHS, an H(2)S donor. In the mice with cerulein-induced acute pancreatitis, the referred hyperalgesia was suppressed by NNC 55-0396 (NNC), a selective T-channel inhibitor; zinc chloride; or ascorbic acid, known to inhibit Ca(v)3.2 selectively among three T-channel isoforms; and knockdown of Ca(v)3.2. In contrast, AP18 and knockdown of TRPA1 had no significant effect on the cerulein-induced referred hyperalgesia, although they significantly potentiated the antihyperalgesic effect of NNC at a subeffective dose. TRPA1 but not Ca(v)3.2 in the dorsal root ganglia was downregulated at a protein level in mice with cerulein-induced pancreatitis. The data indicate that TRPA1 and Ca(v)3.2 mediate the exogenous H(2)S-induced pancreatic nociception in naïve mice and suggest that, in the mice with pancreatitis, Ca(v)3.2 targeted by H(2)S primarily participates in the pancreatic pain, whereas TRPA1 is downregulated and plays a secondary role in pancreatic nociceptive signaling.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido L-ascórbico, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Ácido L-ascórbico, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Ácido L-ascórbico, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Ácido L-ascórbico, 99%
Sigma-Aldrich
Ácido L-ascórbico, reagent grade, crystalline
Supelco
Ácido L-ascórbico, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ácido L-ascórbico, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ácido L-ascórbico, ACS reagent, ≥99%
Sigma-Aldrich
Urethane, ≥99%
Supelco
Ácido L-ascórbico, analytical standard
Sigma-Aldrich
Ácido L-ascórbico, meets USP testing specifications
Sigma-Aldrich
Ácido L-ascórbico, reagent grade
Sigma-Aldrich
Ácido L-ascórbico, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Ácido L-ascórbico, FCC, FG
Sigma-Aldrich
Urethane, ≥99.0% (GC)
Sigma-Aldrich
Ácido L-ascórbico, BioUltra, ≥99.5% (RT)
Ácido L-ascórbico, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ácido L-ascórbico, puriss. p.a., ≥99.0% (RT)
Supelco
Ácido L-ascórbico, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ácido L-ascórbico, tested according to Ph. Eur.
Supelco
Urethane, analytical standard
Sigma-Aldrich
NNC 55-0396 hydrate, ≥98% (HPLC)