MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Formal recycling of e-waste leads to increased exposure to toxic metals: an occupational exposure study from Sweden.
Environment international (2014-10-11)
Anneli Julander, Lennart Lundgren, Lizbet Skare, Margaretha Grandér, Brita Palm, Marie Vahter, Carola Lidén
RESUMEN
Electrical and electronic waste (e-waste) contains multiple toxic metals. However, there is currently a lack of exposure data for metals on workers in formal recycling plants. The objective of this study was to evaluate workers' exposure to metals, using biomarkers of exposure in combination with monitoring of personal air exposure. We assessed exposure to 20 potentially toxic metals among 55 recycling workers and 10 office workers at three formal e-waste recycling plants in Sweden. Workers at two of the plants were followed-up after 6 months. We collected the inhalable fraction and OFC (37-mm) fraction of particles, using personal samplers, as well as spot samples of blood and urine. We measured metal concentrations in whole blood, plasma, urine, and air filters using inductively coupled plasma-mass spectrometry following acid digestion. The air sampling indicated greater airborne exposure, 10 to 30 times higher, to most metals among the recycling workers handling e-waste than among the office workers. The exposure biomarkers showed significantly higher concentrations of chromium, cobalt, indium, lead, and mercury in blood, urine, and/or plasma of the recycling workers, compared with the office workers. Concentrations of antimony, indium, lead, mercury, and vanadium showed close to linear associations between the inhalable particle fraction and blood, plasma, or urine. In conclusion, our study of formal e-waste recycling shows that workers performing recycling tasks are exposed to multiple toxic metals.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido clorhídrico, ACS reagent, 37%
Sigma-Aldrich
Ácido nítrico, ACS reagent, 70%
Sigma-Aldrich
Ácido clorhídrico, ACS reagent, 37%
Sigma-Aldrich
Cloruro de hidrógeno solution, 4.0 M in dioxane
Sigma-Aldrich
Ácido nítrico, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Ácido clorhídrico solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ácido clorhídrico, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Ácido clorhídrico, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Cloruro de hidrógeno solution, 2.0 M in diethyl ether
Sigma-Aldrich
Ácido nítrico, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Ácido clorhídrico, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Ácido clorhídrico, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Ácido clorhídrico solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Ácido nítrico, red, fuming, HNO3 >90 %
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Ácido nítrico, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥65%
Sigma-Aldrich
Ácido clorhídrico, SAJ first grade, 35.0-37.0%
Sigma-Aldrich
Cloruro de hidrógeno solution, 1.0 M in diethyl ether
Sigma-Aldrich
Ácido clorhídrico, JIS special grade, 35.0-37.0%
Sigma-Aldrich
Ácido clorhídrico solution, 1 M
Sigma-Aldrich
Ácido clorhídrico solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Ácido nítrico, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Ácido nítrico solution, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Ácido clorhídrico solution, 6 M
Sigma-Aldrich
Cloruro de hidrógeno solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Ácido nítrico, ACS reagent, ≥90.0%
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Ácido clorhídrico solution, 12 M
Sigma-Aldrich
Ácido clorhídrico solution, 32 wt. % in H2O, FCC