MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • In-depth analysis of site-specific N-glycosylation in vitronectin from human plasma by tandem mass spectrometry with immunoprecipitation.
In-depth analysis of site-specific N-glycosylation in vitronectin from human plasma by tandem mass spectrometry with immunoprecipitation.
Analytical and bioanalytical chemistry (2014-11-07)
Heeyoun Hwang, Ju Yeon Lee, Hyun Kyoung Lee, Gun Wook Park, Hoi Keun Jeong, Myeong Hee Moon, Jin Young Kim, Jong Shin Yoo
RESUMEN
The characterization of site-specific microheterogeneity in glycoprotein is very important for understanding cell biology and disease processes. Vitronectin is well known to be a multifunctional glycoprotein in the blood and the extracellular matrix, which is related to hepatocellular carcinoma (HCC). Here, we systematically analyzed the site-specific N-glycopeptides of vitronectin in human plasma by tandem mass spectrometry combined with immunoprecipitation and hydrophilic interaction liquid chromatography (HILIC) enrichment. Vitronectin was purified with immunoprecipitation by monoclonal antibody from plasma and digested to tryptic N-glycopeptides.Then, enrichment with HILIC materials was used and followed by analysis with nano-LC/MS/MS. The sequences of N-glycopeptides were identified from the mass spectra by high-energy C-trap dissociation (HCD) and collision-induced dissociation (CID). In HCD mode, oxonium ions were used for recognizing glycopeptides and y ions for sequencing the peptide backbone. In CID mode, Y ions were used for characterizing their glycoforms. As a result, a total of 17 site-specific N-glycopeptides were completely identified in all of the three N-glycosylation sites of vitronectin in human plasma, including 12 N-glycopeptides first reported. Finally, we specifically found that three hybrid and four complex glycopeptides of triantennary forms with outer fucosylation increased in HCC human plasma.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido fórmico, reagent grade, ≥95%
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥96%
Sigma-Aldrich
Ácido fórmico, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Yodoacetamida, BioUltra
Sigma-Aldrich
DL-Ditiotreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Yodoacetamida, Single use vial of 56 mg
Sigma-Aldrich
Ácido fórmico, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Yodoacetamida, ≥99% (NMR), crystalline
Supelco
DL-Ditiotreitol solution, 1 M in H2O
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥88%
Sigma-Aldrich
3-Indoleacetic acid, 98%
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
Indole-3-acetic acid sodium salt, suitable for plant cell culture, BioReagent, ≥98%
Sigma-Aldrich
Ácido fórmico, ≥95%, FCC, FG
SAFC
Yodoacetamida
Sigma-Aldrich
Ácido fórmico, JIS special grade, ≥98.0%
Supelco
3-Indoleacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
3-Indoleacetic acid, SAJ special grade, ≥98.5%