MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
NMR-based urinalysis for beta-ketothiolase deficiency.
Clinica chimica acta; international journal of clinical chemistry (2014-09-10)
Chun-Yiu Law, Ching-Wan Lam, Chor-kwan Ching, Kin-Cheong Eric Yau, Tsz-wai Ho, Chi-kong Lai, Chloe Miu Mak
RESUMEN
Beta-ketothiolase deficiency is a rare inborn errors of metabolism (IEM) affecting the catabolism of isoleucine, characterized by severe ketoacidosis in children of 6 to 24months old. A prompt diagnosis is of paramount importance as the metabolic decompensation can be effectively reverted by glucose infusion and health outcomes are improved on a protein-restricted diet. Currently, majority of the laboratory diagnosis were made based on mass-spectrometry and molecular genetics while little is mentioned on the advancement of nuclear magnetic resonance (NMR) spectroscopy for the diagnosis of this condition. We report a case of beta-ketothiolase deficiency in a 1-y-old Chinese boy who presented with repeated vomiting, impaired consciousness and severe ketoacidosis. NMR urinalysis detected excessive amount of butanone (a disease specific marker of beta-ketothiolase deficiency), tiglylglycine, (intermediate of isoleucine catabolism) and ketones. Diagnosis of beta-ketothiolase deficiency was further established by molecular genetic studies of ACAT1 gene of the proband. This case illustrated that NMR-based urinalysis is complementary to organic acid analysis for diagnosis of beta-ketothiolase deficiency. The operation of NMR is simple and fast; sample preparation is a two-step procedure while the NMR acquisition is automatic and usually takes <15min. We envisage that NMR analysis will become more available in clinical laboratories and will play an important role in acute pediatric care.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ≥98%
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, BioXtra, ≥98.0% (titration)