MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Toxic effects of chronic low-dose exposure of thioacetamide on rats based on NMR metabolic profiling.
Journal of pharmaceutical and biomedical analysis (2014-07-06)
Dan-Dan Wei, Jun-Song Wang, Peng-Ran Wang, Ming-Hui Li, Ming-Hua Yang, Ling-Yi Kong
RESUMEN
Thioacetamide (TAA) is a well-known toxicant and its long term exposure could induce liver fibrosis and cirrhosis. A liver fibrosis rat model was established by consecutive injection of TAA solution for 7 weeks. Serum and urine samples were collected weekly for NMR based metabolomic study. Clinical biochemistry of serum samples revealed liver impairment and fibrosis. Histopathological inspections disclosed severe liver fibrosis and cirrhosis formation, and pathological changes in kidney by long-term TAA administration. Orthogonal partial least squares-discriminant analysis (OPLS-DA) was applied on serum and urine samples to excavate differential metabolites associated with TAA induced impairment and explore the time-dependent metabolic event associated with this xenobiotic perturbation. Integration of metabolomics results with serum biochemical revealed several potential biomarkers for liver fibrosis (2-hydroxybutyrate, 3-hydroxybutyrate and adipate in urine, and phenylalanine, N,N-dimethyl glycine, O-acetyl glycoprotein, N-acetyl glycoprotein and choline in serum). Pathway analysis revealed disturbed pathways concerning tricarboxylic acid (TCA) cycle, pyruvate metabolism, starch and sucrose metabolism, glycolysis or gluconeogenesis, degradation of ketone bodies, butanoate metabolism, and biosynthesis of BCAAs (valine, leucine and isoleucine) and AAAs (phenylalanine, tyrosine and tryptophan). This integrative study should help to develop a systematic understanding of liver fibrosis-related diseases and their metabolic events.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Formaldehído solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehído solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehído solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehído solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Thioacetamide, ACS reagent, ≥99.0%
Sigma-Aldrich
Formaldehído solution, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehído solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Thioacetamide, reagent grade, 98%
Sigma-Aldrich
Formaldehído solution, tested according to Ph. Eur.
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Formaldehído solution, 10%
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formaldehído solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehído solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C