MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Direct production of OH radicals upon CH overtone activation of (CH3)2COO Criegee intermediates.
The Journal of chemical physics (2014-12-22)
Fang Liu, Joseph M Beames, Marsha I Lester
RESUMEN
Ozonolysis of alkenes, a principle non-photolytic source of atmospheric OH radicals, proceeds through unimolecular decay of energized carbonyl oxide intermediates, known as Criegee intermediates. In this work, cold dimethyl-substituted Criegee intermediates are vibrationally activated in the CH stretch overtone region to drive the 1,4 hydrogen transfer reaction that leads to OH radical products. IR excitation of (CH3)2COO reveals the vibrational states with sufficient oscillator strength, coupling to the reaction coordinate, and energy to surmount the effective barrier (≤ 16.0 kcal mol(-1)) to reaction. Insight on the dissociation dynamics is gleaned from homogeneous broadening of the spectral features, indicative of rapid intramolecular vibrational energy redistribution and/or reaction, as well as the quantum state distribution of the OH X(2)Π (v = 0) products. The experimental results are compared with complementary electronic structure calculations, which provide the IR absorption spectrum and geometric changes along the intrinsic reaction coordinate. Additional theoretical analysis reveals the vibrational modes and couplings that permit (CH3)2COO to access to the transition state region for reaction. The experimental and theoretical results are compared with an analogous recent study of the IR activation of syn-CH3CHOO and its unimolecular decay to OH products [F. Liu, J. M. Beames, A. S. Petit, A. B. McCoy, and M. I. Lester, Science 345, 1596 (2014)].
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Acetona, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetona, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetona, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Yodo, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
Acetona, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetona, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetona, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Yodo, flakes, ReagentPlus®, ≥99%
Supelco
Yodo, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
Acetona, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%
Supelco
Acetona, analytical standard
Sigma-Aldrich
Hydrazine hydrate, reagent grade, N2H4 50-60 %
Sigma-Aldrich
Hydrazine hydrate solution, puriss. p.a., 24-26% in H2O (RT)
Sigma-Aldrich
Yodo, ≥99.99% trace metals basis
Sigma-Aldrich
Yodo, 99.999% trace metals basis
Sigma-Aldrich
Acetona, histological grade, ≥99.5%
Sigma-Aldrich
Acetona, JIS special grade, ≥99.5%
USP
Acetona, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetona, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Yodo, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Acetona, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetona, SAJ first grade, ≥99.0%
Sigma-Aldrich
Yodo, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Acetona, ≥99%, FCC, FG
Sigma-Aldrich
Yodo, puriss., ≥99.5% (RT), particles (round)
Sigma-Aldrich
Yodo, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Acetona, natural, ≥97%
Sigma-Aldrich
Acetona, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Hydrazine monohydrate, SAJ first grade, ≥98.0%