MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Establishment of an antibody avidity test to differentiate vaccinated cattle from those naturally infected with Mycoplasma bovis.
Establishment of an antibody avidity test to differentiate vaccinated cattle from those naturally infected with Mycoplasma bovis.
Veterinary journal (London, England : 1997) (2014-12-04)
Xiaoxiao Han, Farhan Anwar Khan, Xifang Zhu, Rui Zhang, Riaz Mustafa, Changmin Hu, Yingyu Chen, Huanchun Chen, Aizhen Guo
RESUMEN
Mycoplasma bovis is a major pathogen of bovine respiratory disease (BRD) in China and a live attenuated vaccine has recently been developed. This study aimed to establish an IgG avidity test to differentiate between naturally infected and vaccinated animals. An indirect ELISA (iELISA) was first established in the laboratory to detect antibodies specific to M. bovis using whole cell proteins as coating antigens and serum samples from experimentally infected cattle. The specificity and sensitivity of the iELISA was confirmed using a commercial ELISA kit as a reference standard. Both tests showed substantial agreement as indicated by a κ value of 0.78 (95% confidence interval, CI, 0.62, 0.93), and an overall 92.0% (80/87) agreement between the two tests. Based on the laboratory iELISA, a sodium thiocyanate (NaSCN) competitive iELISA was then developed for the detection of IgG avidity, expressed as relative avidity index (AI). Two-hundred and one experimentally immunised and naturally infected animals were used. These comprised 36 immunised calves, 38 negative control calves, 37 naturally infected calves, 87 calves of unknown status, and an additional three immunised calves that were used for a time trial. By testing true positive and negative antisera from either naturally infected or immunised calves, the AI cut-off value was defined as 70.4%. The diagnostic accuracy of the in-house NaSCN competitive iELISA was determined using serum samples collected from the experimental animals. The IgG avidity test demonstrated 96.0% sensitivity (95% CI 80.5%, 99.3%) and 95.8% specificity (95% CI 79.8%, 99.3%), and was successfully established as a valuable first test for differentiating vaccinated animals from those infected with M. bovis. This test may be a useful tool for clarifying the magnitude of M. bovis infection and in assessing the efficacy of vaccination in exposed animal populations.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Piruvato sódico, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Piruvato sódico, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Piruvato sódico, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Piruvato sódico, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Piruvato sódico, BioXtra, ≥99%
Sigma-Aldrich
Piruvato sódico, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Piruvato sódico, SAJ special grade, ≥95.0%