MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Rapid quantitative chiral amphetamines liquid chromatography-tandem mass spectrometry: method in plasma and oral fluid with a cost-effective chiral derivatizing reagent.
Rapid quantitative chiral amphetamines liquid chromatography-tandem mass spectrometry: method in plasma and oral fluid with a cost-effective chiral derivatizing reagent.
Journal of chromatography. A (2014-07-30)
Matthew N Newmeyer, Marta Concheiro, Marilyn A Huestis
RESUMEN
Methamphetamine is a widely abused psychostimulant containing a chiral center. Consumption of over-the-counter and prescription medications may yield positive amphetamines results, but chiral separation of l- and d-methamphetamine and its metabolite amphetamine can help determine whether the source was licit or illicit. We present the first LC-MS/MS method with precolumn derivatization for methamphetamine and amphetamine chiral resolution in plasma and oral fluid collected with the Oral-Eze(®) and Quantisal™ devices. To 0.5mL plasma, 0.75mL Oral-Eze, or 1mL Quantisal specimen racemic d11-methamphetamine and amphetamine internal standards were added, followed by protein precipitation. Samples were centrifuged and supernatants loaded onto pre-conditioned Phenomenex(®) Strata™-XC Polymeric Strong Cation solid phase extraction columns. After washing, analytes were eluted with 5% ammonium hydroxide in methanol. The eluate was evaporated to dryness and reconstituted in water. Derivatization was performed with 1-fluoro-2,4-dinitrophenyl-5-l-alanineamide (Marfey's reagent) and heating at 45°C for 1h. Derivatized enantiomer separations were performed under isocratic conditions (methanol:water, 60:40) with a Phenomenex(®) Kinetex(®) 2.6μm C18 column. Analytes were identified and quantified by two MRM transitions and their ratio on a 3200 QTrap (AB Sciex) mass spectrometer in ESI negative mode. In all three matrices, the method was linear for all enantiomers from 1 to 500μg/L, with imprecision and accuracy of ≤11.3% and 85.3-108%, respectively. Extraction efficiencies ranged from 67.4 to 117% and matrix effects from -17.0 to 468%, with variation always ≤19.1%. Authentic plasma and OF specimens were collected from an IRB-approved study that included controlled Vicks(®) VapoInhaler™ administration. The present method is sensitive, selective, economic and rapid (separations accomplished in <10min), and improves methamphetamine result interpretation.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Metanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetona, ACS reagent, ≥99.5%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Hidróxido de amonio solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ácido acético, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetona, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ácido acético, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetona, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Ácido fórmico, reagent grade, ≥95%
Sigma-Aldrich
Metanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetona, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥96%
Sigma-Aldrich
Hidróxido de amonio solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Ácido fórmico, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Metanol, anhydrous, 99.8%
Sigma-Aldrich
Ácido acético, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Ácido acético solution, suitable for HPLC
Sigma-Aldrich
Ácido acético, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Ácido acético, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Ácido fórmico, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Metanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetona, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetona, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Metanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥88%
Sigma-Aldrich
Hidróxido de amonio solution, puriss., 30-33% NH3 in H2O
Sigma-Aldrich
Metanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%