MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Novel pyrrolopyrimidines and triazolopyrrolopyrimidines carrying a biologically active sulfonamide moieties as anticancer agents.
Novel pyrrolopyrimidines and triazolopyrrolopyrimidines carrying a biologically active sulfonamide moieties as anticancer agents.
Acta poloniae pharmaceutica (2015-04-09)
Mostafa M Ghorab, Mansour S Alsaisd, Yassin M Nissan
RESUMEN
A new series of pyrroles 5, 6, pyrrolopyrimidines 8, 11-14, 16-29, triazolo-pyrrolopyrimidines 9, 10 and 15 carrying a biologically active sulfonamide moities were synthesized using 2-amino-3-cyano-4-(4-bromophenyl)pyrrole 5 as a strategic starting material. The structures of the prepared compounds were confirmed by elemental analyses, IR, 1H-NMR and 13C-NMR data. All of the synthesized compounds showed promising anticancer activity against breast cancer cell line (MCF7) compared to doxorubicin as reference drug, especially compounds 5-17, 21-24 and 28 with better IC50 than that of doxorubicin. In order to suggest the mechanism of action of their cytotoxic activities, molecular docking on the active site of c-Src was done and good results were obtained.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dimetil sulfóxido-d6, 99.9 atom % D
Sigma-Aldrich
Yodo, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
Dimetil sulfóxido-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimetil sulfóxido-d6, 99.5 atom % D
Sigma-Aldrich
Yodo, flakes, ReagentPlus®, ≥99%
Supelco
Yodo, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
Yodo, ≥99.99% trace metals basis
Sigma-Aldrich
Yodo, 99.999% trace metals basis
Sigma-Aldrich
Dimetil sulfóxido-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Yodo, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Dimetil sulfóxido-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimetil sulfóxido-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Yodo, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Yodo, puriss., ≥99.5% (RT), particles (round)
Sigma-Aldrich
Yodo, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Dimetil sulfóxido-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Dimetil sulfóxido-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS