MilliporeSigma
The human CILP gene: exon/intron organization and chromosomal mapping.
Matrix biology : journal of the International Society for Matrix Biology (1999-12-22)
P Lorenzo, P Aman, Y Sommarin, D Heinegård
RESUMEN
The human cDNA for cartilage intermediate layer protein (CILP) codes for a larger precursor protein that consists of CILP and a homologue to porcine Nucleotide pyrophosphohydrolase (NTPPHase) [Lorenzo et al. 1998a. J. Biol. Chem. 273, 23469-23475]. The human gene has now been isolated and characterized. Southern blot analysis indicated a single copy of the CILP gene in the human genome. The gene spans approximately 15.3 kbp of genomic DNA, and is organized in nine exons. The 5' flanking region contains a putative promoter region with a TATA-like box localized from -29 to -23 bp upstream of the transcription start site. Analysis of the putative promoter region revealed potentially cis-regulatory eukaryotic elements such as GATA-1, MyoD, MZF1, and CdxA. The protein coding region begins in exon 2 with the putative signal peptide. CILP is encoded from exon 3 to exon 9. In addition, exon 9 also codes for the entire NTPPHase homologue and contains the 3' untranslated region of the gene. All the introns follow the 'gt-ag' rule, except the last intron, intron 8, that belongs to the minor class of pre-mRNA introns that contain 'at-ac' at their 5' and 3' ends, respectively. The CILP gene was mapped to human chromosome 15q22.