MilliporeSigma
The structural organization of the human aldehyde reductase gene, AKR1A1, and mapping to chromosome 1p33-->p32.
Cytogenetics and cell genetics (1999-07-07)
J Fujii, R Hamaoka, A Matsumoto, T Fujii, Y Yamaguchi, M Egashira, O Miyoshi, N Niikawa, N Taniguchi
RESUMEN
Genomic DNA encoding for human aldehyde reductase (AKR1A1), a member of the aldo-keto reductase superfamily, was isolated and characterized. The genomic DNA is approximately 16 kb in length and contains eight exons which encode the entire coding region and the 3'-untranslated sequences. AKR1A1 was localized on chromosome 1p33-->p32 by fluorescence in situ hybridization.