MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
A Simple and Efficient Preparation of High-Purity Hydrogen Trioxide (HOOOH).
Angewandte Chemie (International ed. in English) (2015-08-04)
Gregor Strle, Janez Cerkovnik
RESUMEN
A simple and efficient method allows the synthesis of solutions of high-purity hydrogen trioxide (HOOOH), released in the low-temperature methytrioxorhenium(VII) (MTO)-catalyzed transformation of the ozonized polystyrene-supported dimethylphenylsilane. High-purity hydrogen trioxide solutions in diethyl ether, separated from the polymer and free of any reactants and by-products, can be stored at -20 °C for weeks. By removing the solvent in vacuo, HOOOH could be isolated in highly pure form or transferred to other solvents, thus significantly extending the research perspectives of HOOOH for novel applications.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Divinylbenzene, technical grade, 55%
Sigma-Aldrich
Divinylbenzene, technical grade, 80%
Sigma-Aldrich
Acetona, JIS special grade, ≥99.5%
Sigma-Aldrich
Chlorodimethylsilane, 98%
Sigma-Aldrich
Acetona, SAJ first grade, ≥99.0%
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Acetona, ≥99%, FCC, FG
Sigma-Aldrich
Acetona, natural, ≥97%
Sigma-Aldrich
Acetona, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetona, suitable for residue analysis, ≥99.5%
Sigma-Aldrich
Acetona, suitable for HPLC
Sigma-Aldrich
Acetona, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Acetona, suitable for chromatography, ≥99.8%
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
Acetona, suitable for residue analysis, JIS 5000