MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
A-kinase anchoring protein 79/150 coordinates metabotropic glutamate receptor sensitization of peripheral sensory neurons.
Pain (2015-07-15)
Kalina Szteyn, Matthew P Rowan, Ruben Gomez, Junhui Du, Susan M Carlton, Nathaniel A Jeske
RESUMEN
Glutamate serves as the primary excitatory neurotransmitter in the nervous system. Previous studies have identified a role for glutamate and group I metabotropic receptors as targets for study in peripheral inflammatory pain. However, the coordination of signaling events that transpire from receptor activation to afferent neuronal sensitization has not been explored. Herein, we identify that scaffolding protein A-kinase anchoring protein 79/150 (AKAP150) coordinates increased peripheral thermal sensitivity after group I metabotropic receptor (mGluR5) activation. In both acute and persistent models of thermal somatosensory behavior, we report that mGluR5 sensitization requires AKAP150 expression. Furthermore, electrophysiological approaches designed to record afferent neuronal activity reveal that mGluR5 sensitization also requires functional AKAP150 expression. In dissociated primary afferent neurons, mGluR5 activation increases TRPV1 responses in an AKAP-dependent manner through a mechanism that induces AKAP association with TRPV1. Experimental results presented herein identify a mechanism of receptor-driven scaffolding association with ion channel targets. Importantly, this mechanism could prove significant in the search for therapeutic targets that repress episodes of acute pain from becoming chronic in nature.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
L-Glutamina, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
L-Glutamina, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
5-Fluoro-2′-desoxiuridina, thymidylate synthase inhibitor
SAFC
L-Glutamina
Sigma-Aldrich
Capsaicin, natural
Sigma-Aldrich
L-Glutamina, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Pyridine hydrochloride, 98%
Sigma-Aldrich
L-Glutamina, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamina
Sigma-Aldrich
Capsaicin, from Capsicum sp., ≥50% (HPLC)
Sigma-Aldrich
Fenobam, ≥98% (HPLC), solid