MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effect of an Experimental Paste with Hydroxyapatite Nanoparticles and Fluoride on Dental Demineralisation and Remineralisation in situ.
Effect of an Experimental Paste with Hydroxyapatite Nanoparticles and Fluoride on Dental Demineralisation and Remineralisation in situ.
Caries research (2015-08-19)
Beatriz M Souza, Livia P Comar, Mariele Vertuan, Constantino Fernandes Neto, Marília Afonso Rabelo Buzalaf, Ana Carolina Magalhães
RESUMEN
This study evaluated the effect of an experimental paste containing hydroxyapatite in nanoparticles (nano-HA)/fluoride on dental de-remineralisation in situ. Thirteen subjects took part in this crossover/randomised/double-blind study performed in 4 phases (14 days each). Four sound and 4 pre-demineralised specimens were worn intraorally at each phase corresponding to the following treatments: Nanop Plus (10% HA, 0.2% NaF, nano-HA/fluoride), MI Paste Plus (casein phosphopeptide-amorphous calcium phosphate, 0.2% NaF), F (0.2% NaF) and placebo. Two-hundred and forty enamel and 240 dentine specimens were selected by using surface microhardness; half of them were subjected to pre-demineralisation and the other half remained sound. Sound specimens were further exposed to severe cariogenic challenge (20% sucrose in biofilm) in situ, while pre-demineralised specimens were not. All specimens were exposed to fluoride dentifrice slurry 2×1 min/day. Thereafter, the treatments were done for 4 min. The de-remineralisation was quantified by transversal microradiography. The data were statistically analysed by repeated-measures ANOVA/Tukey's tests (p<0.05). Generally, no huge differences were found among the treatments. However, Nanop Plus was the only treatment able to significantly reduce dentine demineralisation (x0394;Z, integrated mineral loss) and to improve enamel remineralisation (x0394;x0394;Z, integrated mineral uptake) compared to placebo. No treatments were able to reduce enamel demineralisation, while for dentine remineralisation all treatments were similarly effective in improving x0394;x0394;Z compared to placebo. Nanop Plus seems to have a positive influence on dental de-remineralisation, which should be further confirmed.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Hidróxido de potasio, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Hidróxido de potasio, reagent grade, 90%, flakes
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Hidróxido de potasio, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Hidróxido de potasio, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Thymol, ≥98.5%
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
Lactic acid, 88%, FCC
Sigma-Aldrich
Hidróxido de potasio, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%
Sigma-Aldrich
Hidróxido de potasio, technical, ≥85%, powder
Sigma-Aldrich
Hidróxido de potasio, ≥99.95% trace metals basis
Sigma-Aldrich
Hidróxido de potasio, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Thymol, FCC, FG
Sigma-Aldrich
Hidróxido de potasio, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
Thymol, meets analytical specification of Ph. Eur., BP, NF, 99-101%
Sigma-Aldrich
Hidróxido de potasio, puriss. p.a., ≥86% (T), pellets
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium Fluoride Solution