MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Potentiation of cytokine-induced proliferation of human Natural Killer cells by intravenous immunoglobulin G.
Clinical immunology (Orlando, Fla.) (2015-08-27)
Andrew C Issekutz, Beata Derfalvi, Fabian Käsermann, Derek Rowter
RESUMEN
Intravenous IgG (IVIG) therapy can be used for immunomodulation. IL-2 is an immunoregulatory cytokine. We evaluated IVIG modulation of human blood lymphocyte response to IL-2 and other cytokines. Neither IVIG nor low concentrations of IL-2 (3-30U/ml) induced lymphocyte proliferation, but in combination they synergistically enhanced proliferation of NK cells. The CD56(bright) cells expanded more than CD56(dim) NK cells, with 90% of NK cells dividing up to 8 generations by day 6, while <8% of T cells divided. IVIG also potentiated NK cell proliferation with IL-12, IL-15 and IL-18. The IVIG+cytokine-expanded NK cells were less cytotoxic for K562 cells, than NK cells with cytokine alone. IVIG also enhanced interferon-γ production with IL-2, IL-12 and IL-18. In conclusion, IVIG selectively potentiates NK cell proliferation and interferon-γ secretion with IL-2, IL-12, IL-15 and IL-18 in vitro. These findings warrant evaluation in vivo in relation to NK cells and the immunoregulatory actions of IVIG.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Timidina, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Timidina, ≥99%
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
Timidina, ≥99.0% (HPLC)
Sigma-Aldrich
Interleukin-2, human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
IL-2 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, carrier free
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL