MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
The induction of pro-angiogenic processes within a collagen scaffold via exogenous estradiol and endometrial epithelial cells.
Biotechnology and bioengineering (2015-05-07)
Jacquelyn C Pence, Kathryn B H Clancy, Brendan A C Harley
RESUMEN
Nutrient transport remains a major limitation in the design of biomaterials. One approach to overcome this constraint is to incorporate features to induce angiogenesis-mediated microvasculature formation. Angiogenesis requires a temporal presentation of both pro- and anti-angiogenic factors to achieve stable vasculature, leading to increasingly complex biomaterial design scheme. The endometrium, the lining of the uterus and site of embryo implantation, exemplifies a non-pathological model of rapid growth, shedding, and re-growth of dense vascular networks regulated by the dynamic actions of estradiol and progesterone. In this study, we examined the individual and combined response of endometrial epithelial cells and human umbilical vein endothelial cells to exogenous estradiol within a three-dimensional collagen scaffold. While endothelial cells did not respond to exogenous estradiol, estradiol directly stimulated endometrial epithelial cell transduction pathways and resulted in dose-dependent increases in endogenous VEGF production. Co-culture experiments using conditioned media demonstrated estradiol stimulation of endometrial epithelial cells can induce functional changes in endothelial cells within the collagen biomaterial. We also report the effect of direct endometrial epithelial and endothelial co-culture as well as covalent immobilization of estradiol within the collagen biomaterial. These efforts establish the suitability of an endometrial-inspired model for promoting pro-angiogenic events within regenerative medicine applications. These results also suggest the potential for developing biomaterial-based models of the endometrium.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Formaldehído solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehído solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
L-Glutamina, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
1,2-Dicloroetano, anhydrous, 99.8%
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, commercial grade, powder
Sigma-Aldrich
L-Glutamina, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, crystalline
SAFC
L-Glutamina
Sigma-Aldrich
Formaldehído solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, BioXtra
Sigma-Aldrich
Ácido acético, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Formaldehído solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
β-Estradiol, meets USP testing specifications
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
L-Glutamina, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ácido acético, ≥99.7%
Sigma-Aldrich
Ácido acético, ≥99.5%, FCC, FG
Sigma-Aldrich
Ácido acético, natural, ≥99.5%, FG
Sigma-Aldrich
Ácido acético, JIS special grade, ≥99.7%
Sigma-Aldrich
Ácido acético solution, 1 N, 1 M
Sigma-Aldrich
L-Glutamina, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamina
Sigma-Aldrich
Ácido acético, SAJ first grade, ≥99.0%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Formaldehído solution, 10%
Sigma-Aldrich
Formaldehído solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehído solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer