MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Carbon dots derived from rose flowers for tetracycline sensing.
Talanta (2015-06-07)
Yuanjiao Feng, Dan Zhong, Hong Miao, Xiaoming Yang
RESUMEN
Herein, an innovative and simple method for synthesizing carbon dots (CDs) with satisfactory fluorescence has been successfully established while rose flowers served as carbon source for the first time. Meanwhile, the fluorescence (FL) mechanism of current CDs was elucidated in detail by fluorescence, UV-vis, HR-TEM, and FTIR-based analyses. Subsequently, this type of CDs was employed for detecting tetracycline (TC) on the basis of the interactions between TC and CDs, and allowed quenching their fluorescence. Moreover, the proposed analytical strategy permitted detecting TC in a linear range of 1.0×10(-8)-1.0×10(-4) mol/L with a detection limit of 3.3×10(-9) mol/L at a signal-to-noise ratio of 3. Significantly, the CDs described here were further applied for fluorescent coding, demonstrating their promising future towards various applications in analytic science.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
L-Glutatión reducido, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutatión reducido, ≥98.0%
Sigma-Aldrich
4-Aminobenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Phosphorus pentoxide, ReagentPlus®, 99%
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
4-Aminobenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%
Sigma-Aldrich
L-Glutatión reducido, BioXtra, ≥98.0%
Supelco
Ibuprofen
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Phosphorus pentoxide, desiccant, with moisture indicator
Sigma-Aldrich
4-Aminobenzoic acid, purified by sublimation, ≥99%
Supelco
Phosphorus pentoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus pentoxide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)