MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Sequential Intracorneal Ring Segment Implantation and Corneal Transepithelial Collagen Cross-Linking in Keratoconus.
Cornea (2015-09-12)
Hany S Elsaftawy, Mansour Hassan Ahmed, Mohamed Yasser Sayed Saif, Rasha Mousa
RESUMEN
To assess the clinical outcomes after sequential surgical implantation of intracorneal ring segments and transepithelial corneal collagen cross-linking (TE-CXL) in keratoconic eyes. In this consecutive randomized study, 40 eyes of 29 patients with progressive mild to moderate keratoconus were included and were randomly divided into two groups. Group 1 included 20 eyes that underwent Keraring implantation, and group 2 included 20 eyes that underwent Keraring insertion followed by TE-CXL 1 month later. In both groups, channel creation was performed manually. Visual, refractive, and topographic outcomes were measured preoperatively and 1, 3, and 6 months postoperatively. There was statistically significant improvement in both groups regarding uncorrected distance visual acuity (P < 0.001) and corrected distance visual acuity (P < 0.001), with a significant reduction in the refractive error and keratometric values (P < 0.001). There was no statistically significant difference between both groups regarding changes in uncorrected distance visual acuity, corrected distance visual acuity, and refractive error (P > 0.05). However, group 2 revealed a more statistically significant reduction in spherical refraction after 3 months (P = 0.04) and highly significant improvement after 6 months (P = 0.004). Intracorneal ring segment implantation is an effective procedure for visual and refractive improvement in keratoconic eyes. The addition of TE-CXL to the procedure compensates for residual superficial irregularities, disrupts the progression of the disease, and provides greater improvements in spherical values. The process is simply a deep stretch with superficial lamellar stiffness.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cloruro de sodio, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
(−)-Riboflavina, from Eremothecium ashbyii, ≥98%
Sigma-Aldrich
Cloruro de sodio, JIS special grade, ≥99.5%
Supelco
Riboflavina (B2), analytical standard
Sigma-Aldrich
(−)-Riboflavina, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Cloruro de sodio, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Diclofenac sodium salt
Sigma-Aldrich
Cloruro de sodio, 99.999% trace metals basis
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
(−)-Riboflavina, meets USP testing specifications
Sigma-Aldrich
Cloruro de sodio, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Cloruro de sodio, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Cloruro de sodio, tablet
Sigma-Aldrich
Cloruro de sodio, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Cloruro de sodio, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.1 M