MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Modulation of Macrophage Functional Polarity towards Anti-Inflammatory Phenotype with Plasmid DNA Delivery in CD44 Targeting Hyaluronic Acid Nanoparticles.
Modulation of Macrophage Functional Polarity towards Anti-Inflammatory Phenotype with Plasmid DNA Delivery in CD44 Targeting Hyaluronic Acid Nanoparticles.
Scientific reports (2015-11-19)
Thanh-Huyen Tran, Ruchir Rastogi, Juili Shelke, Mansoor M Amiji
RESUMEN
The purpose of this study was to modulate macrophage polarity from the pro-inflammatory M1 to anti-inflammatory M2 phenotype using plasmid DNA (pDNA) expressing interleukin-4 (IL4) or interleukin-10 (IL10)-encapsulated in hyaluronic acid-poly(ethyleneimine) (HA-PEI) nanoparticles (NPs). The HA-PEI/pDNA NPs with spherical shape, average size of 186 nm were efficiently internalized by J774A.1 macrophages. Transfection of HA-PEI/pDNA-IL4 and HA-PEI/pDNA-IL10 NPs increased IL4 and IL10 gene expression in J774 macrophages which could re-program the macrophages from M1 to M2 phenotype as evidenced by a significant increase in the Arg/iNOS level, and upregulation of CD206 and CD163 compared to untreated macrophages. Following intraperitoneal (IP) injection to C57BL/6 mice, HA-PEI NPs effectively targeted peritoneal macrophages over-expressing CD44 receptor. In an in vivo model of stimulated peritoneal macrophages, IP administration of HA-PEI/pDNA-IL4 and HA-PEI/pDNA-IL10 to C57BL/6 mice significantly increased the Arg/iNOS ratio and CD163 expression in the cells. Furthermore, HA-PEI/pDNA-IL10 NPs significantly increased peritoneal and serum IL10 levels which effectively suppressed LPS-induced inflammation by reducing level of TNF-α and IL-1β in peritoneal macrophages and in the peritoneal fluid. The results demonstrated that pDNA-IL10-encapsulate HA-PEI NPs skewed macrophage functional polarity from M1 toward an anti-inflammatory M2 phenotype which may be a promising platform for the treatment of inflammatory diseases.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
1,2-Dicloroetano, ACS reagent, ≥99.0%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
1,2-Dicloroetano, anhydrous, 99.8%
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, commercial grade, powder
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, crystalline
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, BioXtra
Sigma-Aldrich
1,2-Dicloroetano, JIS special grade, ≥99.5%
Sigma-Aldrich
1,2-Dicloroetano, SAJ first grade, ≥99.0%