MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Proteomics Reveals a Role for Attachment in Monocyte Differentiation into Efficient Proinflammatory Macrophages.
Journal of proteome research (2015-07-29)
Nataliya K Tarasova, A Jimmy Ytterberg, Karin Lundberg, Xing-Mei Zhang, Robert A Harris, Roman A Zubarev
RESUMEN
Monocytes are blood-borne cells of the innate immune system. They can be differentiated and activated into proinflammatory macrophages that might be employed in tumor immune therapy. Monocyte exposure to lipopolysaccharide (LPS) is a standard method to induce a proinflammatory macrophage state, with the resultant population comprising both adherent and nonadherent cells. In the current study, we aimed to identify the differences in proteomes of these monocyte subpopulations, which addresses a more general question about the role of attachment in monocyte differentiation. Label-free proteomics of a model of human monocytes (THP-1 cell line) revealed that the cells remaining in suspension upon LPS treatment were activated by cytokines and primed for rapid responsiveness to pathogens. In terms of proteome change, the adhesion process was orthogonal to activation. Adherent cells exhibited signs of differentiation and enhanced innate immune responsivity, being closer to macrophages. These findings indicate that adherent, LPS-treated cells would be more appropriate for use in tumor therapeutic applications.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dimetilsulfóxido, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimetilsulfóxido, Molecular Biology
Sigma-Aldrich
Dimetilsulfóxido, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimetilsulfóxido, anhydrous, ≥99.9%
Sigma-Aldrich
Forbol 12-miristato 13-acetato, ≥99% (TLC), film or powder
Sigma-Aldrich
Dimetilsulfóxido, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Dimetilsulfóxido, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Dimetilsulfóxido, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimetilsulfóxido, PCR Reagent
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Forbol 12-miristato 13-acetato, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Dimetilsulfóxido, ≥99.5%
Sigma-Aldrich
Dimetilsulfóxido, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimetilsulfóxido, Vetec, reagent grade, 99%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimetilsulfóxido, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimetilsulfóxido, suitable for HPLC
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Dimetilsulfóxido, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimetilsulfóxido, SAJ first grade, ≥99.0%