MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Toxicity and molecular effects of di-n-butyl phthalate (DBP) on CYP1A, SOD, and GPx in Cyprinus carpio (common carp).
Environmental monitoring and assessment (2015-06-13)
Hizlan H Agus, Sibel Sümer, Figen Erkoç
RESUMEN
Di-n-butyl phthalate (DBP), a widely used plasticizer in the plastic industry, affects regulation of the endocrine system and causes toxicity in animals. In the present study, we evaluated a series of ecotoxicological stress biomarkers in the common carp (Cyprinus carpio) as an experimental model to test for alterations in gene expression at a sublethal concentration of 1 mg/L DBP for 4, 24, and 96 h. In gills, an immediate increase in CYP1A messenger RNA (mRNA) levels was observed within the first 4 h and persisted for 96 h. Protein levels were nearly consistent with mRNA levels. However, a time-dependent inhibition was observed in CYP1A levels in the liver within 96 h. Superoxide dismutase (SOD) and glutathione peroxidase (GPx) levels increased gradually in liver with exposure time to a maximum level of 11-fold. Varied responses of different tissues were likely due to xenobiotic metabolism of DBP. In conclusion, evaluating the tissue-specific alterations of CYP1A, SOD, and GPx levels can be used as specific and effective biomarkers for ecotoxicological monitoring of DBP pollution. We strongly recommend using molecular tools to ecotoxicologists for aquatic monitoring of newly emerging pollutants.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Cloruro de sodio, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Cloruro de sodio, JIS special grade, ≥99.5%
Sigma-Aldrich
Cloruro de sodio, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Fosfato de sodio dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Nitroazul de tetrazolio, ≥90.0% (HPLC)
Sigma-Aldrich
Cloruro de sodio, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Cloruro de sodio, 99.999% trace metals basis
Sigma-Aldrich
Acetona, JIS special grade, ≥99.5%
Sigma-Aldrich
Cloruro de sodio, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Nitroazul de tetrazolio, powder, electrophoresis grade
Sigma-Aldrich
Cloruro de sodio, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetona, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetona, ≥99%, FCC, FG
Sigma-Aldrich
Acetona, natural, ≥97%
Sigma-Aldrich
Acetona, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Fosfato de sodio dibasic dodecahydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Cloruro de sodio, tablet
Sigma-Aldrich
Cloruro de sodio, random crystals, 99.9% trace metals basis