MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Total and Differential Phylloquinone (Vitamin K1) Intakes of Preterm Infants from All Sources during the Neonatal Period.
Nutrients (2015-10-02)
Paul Clarke, Simon J Mitchell, Martin J Shearer
RESUMEN
All newborns require phylloquinone after birth to prevent vitamin K deficiency bleeding. Babies born prematurely may be at particular risk of deficiency without adequate supplementation during infancy. The main sources of phylloquinone in preterm babies during the neonatal period are the prophylactic dose of phylloquinone given at birth, and that derived from parenteral and/or enteral feeding. This observational study formed part of a prospective, multicentre, randomised, controlled trial that examined the vitamin K status of preterm infants after random allocation to one of three phylloquinone prophylactic regimens at birth (0.5 or 0.2 mg intramuscularly or 0.2 mg intravenously). In this nutritional sub-study we quantified the proportional and total phylloquinone intakes of preterm infants within the neonatal period from all sources. Almost all infants had average daily phylloquinone intakes that were in excess of the currently recommended amounts. In infants who did not receive parenteral nutrition, the bolus dose of phylloquinone given at birth was the major source of phylloquinone intake, whereas in infants who received parenteral nutrition, the intake from the parenteral preparation exceeded that from the bolus dose by a ratio of approximately 3:1. Our study supports the concern of others that preterm infants who receive current parenteral nutrition formulations may be receiving excessive vitamin K.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
5-Hydroxymethyl-2-furaldehyde, 99%
Sigma-Aldrich
5-(Hydroxymethyl)furfural, ≥99%, FG
Sigma-Aldrich
Vitamin K1, viscous liquid
Sigma-Aldrich
Vitamin K1, BioXtra, ≥99.0% (sum of isomers, HPLC), mixture of isomers
Supelco
Phylloquinone (K1), analytical standard