MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
The intracellular domains of Notch1 and Notch2 are functionally equivalent during development and carcinogenesis.
Development (Cambridge, England) (2015-06-13)
Zhenyi Liu, Eric Brunskill, Barbara Varnum-Finney, Chi Zhang, Andrew Zhang, Patrick Y Jay, Irv Bernstein, Mitsuru Morimoto, Raphael Kopan
RESUMEN
Although Notch1 and Notch2 are closely related paralogs and function through the same canonical signaling pathway, they contribute to different outcomes in some cell and disease contexts. To understand the basis for these differences, we examined in detail mice in which the Notch intracellular domains (N1ICD and N2ICD) were swapped. Our data indicate that strength (defined here as the ultimate number of intracellular domain molecules reaching the nucleus, integrating ligand-mediated release and nuclear translocation) and duration (half-life of NICD-RBPjk-MAML-DNA complexes, integrating cooperativity and stability dependent on shared sequence elements) are the factors that underlie many of the differences between Notch1 and Notch2 in all the contexts we examined, including T-cell development, skin differentiation and carcinogenesis, the inner ear, the lung and the retina. We were able to show that phenotypes in the heart, endothelium, and marginal zone B cells are attributed to haploinsufficiency but not to intracellular domain composition. Tissue-specific differences in NICD stability were most likely caused by alternative scissile bond choices by tissue-specific γ-secretase complexes following the intracellular domain swap. Reinterpretation of clinical findings based on our analyses suggests that differences in outcome segregating with Notch1 or Notch2 are likely to reflect outcomes dependent on the overall strength of Notch signals.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Cloruro de amonio, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Cloruro de amonio, 99.998% trace metals basis
Sigma-Aldrich
Acetona, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Cloruro de amonio, 99.99% trace metals basis
Sigma-Aldrich
Acetona, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetona, ≥99%, FCC, FG
Sigma-Aldrich
Acetona, natural, ≥97%
Sigma-Aldrich
Cloruro de amonio, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Acetona, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetona, suitable for residue analysis, ≥99.5%
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
Acetona, suitable for HPLC
Sigma-Aldrich
Acetona, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Acetona, suitable for chromatography, ≥99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Acetona, suitable for residue analysis, JIS 5000