MilliporeSigma
Identification and Metabolite Profiling of Chemical Activators of Lipid Accumulation in Green Algae.
Plant physiology (2017-06-28)
Nishikant Wase, Boqiang Tu, James W Allen, Paul N Black, Concetta C DiRusso
RESUMEN
Microalgae are proposed as feedstock organisms useful for producing biofuels and coproducts. However, several limitations must be overcome before algae-based production is economically feasible. Among these is the ability to induce lipid accumulation and storage without affecting biomass yield. To overcome this barrier, a chemical genetics approach was employed in which 43,783 compounds were screened against