MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Li-O
The journal of physical chemistry letters (2017-08-22)
Yang Li, Shanmu Dong, Bingbing Chen, Chenglong Lu, Kailiang Liu, Zhonghua Zhang, Huiping Du, Xiaogang Wang, Xiao Chen, Xinhong Zhou, Guanglei Cui
RESUMEN
Redox mediators (RMs) have been widely applied to reduce the charge overpotential of nonaqueous lithium-oxygen (Li-O
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Tetraethylene glycol, 99%