- Anti-inflammatory mechanism of galangin in lipopolysaccharide-stimulated microglia: Critical role of PPAR-γ signaling pathway.
Anti-inflammatory mechanism of galangin in lipopolysaccharide-stimulated microglia: Critical role of PPAR-γ signaling pathway.
Since microglia-associated neuroinflammation plays a pivotal role in the progression of neurodegenerative diseases, controlling microglial activation has been suggested as a potential therapeutic strategy. Here, we investigated the anti-inflammatory effects of galangin (3,5,7-trihydroxyflavone) in microglia and analyzed the underlying molecular mechanisms. Galangin inhibited the expression of inducible nitric oxide synthase (iNOS) and pro-inflammatory cytokines and enhanced the expression of anti-inflammatory interleukin (IL)-10 in lipopolysaccharide (LPS)-stimulated BV2 microglia. Galangin also suppressed microglial activation and the expression of pro-inflammatory markers in LPS-injected mouse brains. The results of mechanistic studies have shown that galangin inhibited LPS-induced phosphorylation of p38 mitogen activated protein kinase (MAPK), c-Jun N-terminal kinase (JNK), phosphatidylinositol 3-kinase (PI3K)/Akt, and nuclear factor (NF)-κB activity. On the contrary, galangin increased the activity of transcription factors, such as nuclear factor-E2-related factor 2 (Nrf2), cAMP response element-binding protein (CREB), and peroxisome proliferator-activated receptor (PPAR)-γ, known to play an anti-inflammatory role. In addition, galangin showed antioxidant effects by suppressing the expression of NADPH oxidase subunits p47