MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Rapid onset pressor response to exercise in young women with a family history of hypertension.
Experimental physiology (2017-07-06)
Evan L Matthews, Jody L Greaney, Megan M Wenner
RESUMEN
What is the central question of this study? Alterations in blood pressure control at exercise onset are apparent in older adults with established cardiovascular disease. It is currently not known whether these alterations are evident in young adults with a family history of hypertension. What is the main finding and its importance? We demonstrate that young women with a family history of hypertension display a larger change in blood pressure within the first 10 s of isometric exercise. These data suggest altered blood pressure control in young women with a family history of hypertension. Hypertensive adults demonstrate atypical increases in blood pressure (BP) and muscle sympathetic nerve activity (MSNA) at the immediate onset of static muscle contraction. However, it is unknown whether these abnormal responses occur in young, otherwise healthy adults at risk for developing future disease, such as those with a family history of hypertension (+FH). We tested the hypothesis that +FH young women have exaggerated increases in BP and MSNA at the onset of static muscle contraction compared with those without a family history of hypertension (-FH). We retrospectively examined beat-by-beat BP and MSNA during the initial 30 s of isometric handgrip exercise (30% of maximal voluntary contraction) in 16 +FH (22 ± 2 years old, 22 ± 3 kg m
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder