Skip to Content
Merck

UPMPCFDNA

Ultra pure MagPrep® cfDNA Isolation Kit

new
greener alternative

No proteinase K or heating step needed for Magprep® Nucleic Acid Isolation,Nuclease free (NICKase, RNase, DNase), DNA-free, High-throughput Compatible and Manual

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

UNSPSC Code:
41105500
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

usage

sufficient for 100 purifications

sustainability

Greener Alternative Product, Greener Alternative Product

concentration

45-55 mg/mL (Magnetic particle)

test parameters

total test time: 5 min

suitability

suitable for nucleic acid purification

greener alternative category

storage temp.

2-30°C

General description

The Ultra pure MagPrep® cfDNA isolation kit is designed for highly efficient cfDNA nucleic acid isolation from most biological fluids in both manual or high-throughput automated platforms. The kit contains magnetic silica particles and uniquely formulated buffers optimized for rapid easy handling and maximal yield. Nucleic acid isolation is achieved without the need for Proteinase K or sample heating, and all items are shipped and stored at ambient temperature. All kit components are quality tested to be free of contaminant nucleases and DNA, reducing the likelihood of false positives or negatives in downstream assays. The purified nucleic acid eluate is free of PCR inhibitors such as chaotropic salts or alcohols, allowing for effective amplification in various downstream applications such as RT-qPCR, ddPCR, NGS, and cDNA generation. For research or further manufacturing use only. Not intended for direct use in humans or animals. Non-TSCA use only in US.
We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of the four categories of greener alternatives. Click here to view its DfS scorecard.

Features and Benefits

  • Nucleic Acid Isolation
  • Manual or high-throughput automated isolations
  • RT-qPCR
  • Digital PCR
  • Biological samples can be used
  • Room temperature stable reagents
  • DNA free
  • DNase free
  • RNase free
  • NICKase free
  • simple and fast protocols

Legal Information

MAGPREP is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Only

Product No.
Description

  • UPMPPART-2ML-KC Ultra pure MagPrep Magnetic Particles 1

  • UPMPCFDBB-50ML-KC Ultra pure MagPrep cfDNA Binding Buffer 1

  • UPMPWB1-100ML-KC Ultra pure MagPrep Wash Buffer 1 1

  • UPMPWB2-100ML-KC Ultra pure MagPrep Wash Buffer 2 1

  • UPMPEB-10ML-KC Ultra pure MagPrep Elution Buffer 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service