MilliporeSigma
Smart Lab.
Smarter Access.
Connected Lab Instrumentation
Five instruments sitting on a laboratory bench with a scientist in a lab coat walking past in the background.
The laboratory of the future requires next-level tools for next-level science. Modern labs can leverage technology that expands access, improves collaboration, standardizes methods, and enables scientists to work more safely and efficiently.  Our connected lab instrumentation offers intuitively-designed, ergonomic tools featuring cloud, Bluetooth® and Wi-Fi® capabilities that support greater productivity and help elevate your research.
Click on each instrument name below for information.
Millicell® ERS 3.0 Digital Voltohmmeter
Photograph of a Millicell ERS 3.0 Digital Voltohmmeter, with standing in-well probe in a 24-well plate of cell culture inserts.
Experience the better way to TEER. The Millicell® ERS 3.0 Digital Voltohmmeter acquires highly stable measurements while automatically preserving your data.
  • Easy-to-use touchscreen interface
  • Standing in-well probe for rapid, steady readings
  • Automatic, on-instrument data logging
  • Plug-in or battery pack power source for use on the bench or in the hood
  • Adjustable electrode, compatible with a wide variety of cell culture inserts
Your TEER data can easily be exported via USB drive, ethernet, or uploaded to the cloud. Our Millicell® Cloud features comprehensive measurement view, enabling visualization and analysis your data in various formats. Your Millicell® ERS 3.0 data is automatically graphed or plotted in the cloud. Filter data by user, project, cell line, or device to view only the desired results. You can also view your plate summary, download your raw data (csv), or archive data as needed.
Related Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Benefits
Streamline analysis
Scientist sitting at bench, looking at Millicell® DCI screen showing confluency mask of cell culture in T-75 flask
    Work smarter
    Counting cells with the Scepter™ 3.0 handheld cell counter
      Expand capabilities
      Scientist sitting at a desk, looking at a computer with data.
        REQUEST INFORMATION
        To speak with a representative about our connected lab instrumentation, please complete the below form and we will be in touch.
        Fields indicated by an * are required.