The better way to TEER
Millicell® ERS 3.0 Digital Voltohmmeter
Discover a better way to TEER
The Millicell® ERS 3.0 streamlines data capture with intuitive, user-friendly enhancements. Achieve more stable measurements using a self-standing, in-well probe. Keep track of your results with real-time, on-instrument voltage or resistance data recording.
Simplify analysis across wells and between experiments using a cloud-based application. Obtain consistent readings with low noise, greater resolution, and high accuracy. Optimize your transepithelial electrical resistance experiments with the Millicell® ERS 3.0.
A new TEER experience
The Millicell® ERS 3.0 offers features for simplified data capture and enhanced analysis.
- Intuitive touchscreen display
- Ergonomic standing in-well probe
- On-instrument data logging and background subtraction
- Option to export results via ethernet or USB drive, or upload to cloud
- Corded power or battery pack power source for use on the bench or in the hood
- Adjustable electrode, compatible with 6-, 12-, and 24-well formats with an accessory probe for 96-well formats
- Built-in sensor to measure and record media temperature
- Additional cloud-based application for convenient downstream analysis and data archiving
- Resistance range of 0 – 100 kΩ, with 1 Ω resolution
Related Products
Response not successful: Received status code 500
Move up a TEER
Using the Millicell® ERS-2? It might be the right time to upgrade your instrument. Learn more about our Millicell® ERS Instrument Upgrade Program.
| Millicell® ERS-2 Voltohmmeter | Millicell® ERS 3.0 Digital Voltohmmeter |
Resistance and voltage measurement | ✓ | ✓ |
Temperature measurement | ✓ | |
Touchscreen display | ✓ | |
Ergonomic self-standing probe | ✓ | |
On-instrument data storage | ✓ | |
Automatic background subtraction | ✓ | |
Cloud-based data storage | ✓ | |
Cloud-based analysis | ✓ | |
Export data to USB drive |
| ✓ |
Optimize your workflow
Monitor Cells
Use the Millicell® DCI Digital Cell Imager for quick, objective, in-vessel determination of common cell culture parameters including confluency, cell count, and morphology.
Collect TEER Data
Move to the Millicell® ERS 3.0 for rapid, reliable TEER data. Analyze your experimental data in the cloud from anywhere. Our Connected Lab Instruments support connectivity for a more holistic view of cell health, monolayer structure, and barrier formation for better analysis.
Analyze Results
Analyze your experimental data in the Millicell® Cloud from anywhere.