MilliporeSigma
Buchwald Catalysts & Ligands
As a chemist, you're focused on discovering new chemistry and its practical applications. We support your breakthroughs with an extensive portfolio of Buchwald catalysts and ligands. In collaboration with Stephen Buchwald and his MIT research group, we offer highly active palladium precatalysts and biaryl phosphine ligands for efficient cross-coupling reactions, forming bonds like C-C, C-N, and more. These electron-rich, tunable ligands provide stable, reactive catalyst systems, reducing catalyst loadings, reaction times, and eliminating the need for reducing agents, enabling new methods not achievable with traditional Pd sources. 
A chemical structure showing a biphenyl backbone with a phosphine group (PR₂) attached to the ortho position of the upper phenyl ring. The lower phenyl ring has two substituents labeled R′ and R″ at the ortho and para positions relative to the biphenyl linkage.
Buchwald Ligands
  • Buchwald and his team developed biaryl phosphine ligands that enhance the reaction’s efficiency, selectivity, and versatility.
  • Earlier ligands like dicyclohexyl phosphine dimethylamine enabled aryl chloride–alkyl amine couplings.
  • Advanced ligands (e.g. XPhos, tBuXPhos) offer higher activity and compatibility with hindered substrates.
  • Bulky ligands enhanced catalytic performance and conversion rates in C–N couplings.
  • The Buchwald group also developed copper-catalyzed N-arylation for imidazoles and other heterocycles.
A chemical structure showing a palladium complex labeled “Gen I.” The central palladium (Pd) atom is bonded to a chloride (Cl), an amine ligand (NH₂) attached through a benzylamine-type ring system, and a generic ligand represented as L. A benzene ring is fused to the benzylamine-type ring system, orthogonally to the palladium.
Buchwald Gen 1 Precatalysts
  • First-generation (G1) precatalysts employ 2-phenylethan-1-amine–based ligands for enhanced stability in both solution and solid phases.
  • Air- and moisture-stable, enabling convenient handling.
  • LPd(0) forms in situ via base-promoted reductive elimination, yielding indoline as a byproduct.
  • Higher temperatures are often used with weaker bases to facilitate ligand deprotonation and activate the catalyst.
A chemical structure showing a palladium complex labeled “Gen II.” The central palladium (Pd) atom is bonded to a chloride (Cl), an amine ligand (NH₂) attached through a benzylamine-type ring system, and a generic ligand represented as L. Two benzene rings are fused to the benzylamine-type ring system, ortho and para to the palladium.
Buchwald Gen 2 Precatalysts
  • Second-generation (G2) precatalysts use 2-aminobiphenyl instead of 2-phenylethan-1-amine.
  • Shows improved reactivity with weaker bases (e.g., phosphates, weak carbonates).
  • Often enable catalyst activation at lower temperatures.
  • Enhanced reactivity arises from the greater acidity of the aromatic amine in G2 vs. the aliphatic amine in G1.
  • Key features: air and moisture stable, high efficiency, mild conditions, short reaction times, and low catalyst loadings.
  • Demonstrated excellent performance in Suzuki cross-coupling reactions.
Two chemical structures showing palladium complexes labeled “Gen III” and “Gen IV.” The central palladium (Pd) atom of each is bonded to a chloride (Cl), an amine ligand (NH₂) attached through a benzylamine-type ring system, a generic ligand represented as L, and a mesylate ligand (OMs). Two benzene rings are fused to the benzylamine-type ring system, ortho and para to the palladium. The Gen IV molecule is distinguished by a methyl group attached to the amine ligand.
Buchwald Gen 3 and 4 Precatalysts
  • G3 and G4 Buchwald precatalysts are advanced, stable palladium complexes for diverse cross-coupling reactions (C–C, C–N, C–O, C–F, C–CF₃, C–S).
  • Offer good solubility and activity, which can allow the use of lower catalyst loadings and/or shorter reaction times.
  • G3 uses a mesylate ligand (vs. chloride) to enhance solubility, stability, and accommodate bulkier ligands.
  • G4 features N-methyl-2-aminobiphenyl as the amine ligand, generating a more benign byproduct (N-methyl carbazole vs carbazole).
  • Both generations are effective in key reactions like Suzuki-Miyaura, aminocarbonylation, and N-arylation.
Chemical structure of a sixth generation palladium complex featuring a palladium (Pd) center bonded to a bromine (Br) atom, a generic ligand represented as L, and a phenyl group with a trifluoromethyl (CF₃) substituent para to the palladium.
Buchwald Gen 6 Precatalysts
  • G6 precatalysts are oxidative addition complexes (OACs), offering advanced performance. These on-cycle precatalysts facilitate easier catalyst activation.
  • Retain thermal and air stability while enabling base-free activation and simplified synthesis.
  • Avoid generation of carbazole byproducts during precatalyst activation.
  • Can support extremely bulky ligands and improve solubility and stability.
  • Enable efficient formation of C–C, C–N, C–O, C–F, and C–S bonds.
  • Consistently deliver higher reactivity and better yields than earlier generations.
Related Resources
Page 1 of 2
References
1.
Biscoe MR, Fors BP, Buchwald SL. 2008. A New Class of Easily Activated Palladium Precatalysts for Facile C−N Cross-Coupling Reactions and the Low Temperature Oxidative Addition of Aryl Chlorides. J. Am. Chem. Soc.. 130(21):6686-6687. https://doi.org/10.1021/ja801137k
2.
Kinzel T, Zhang Y, Buchwald SL. 2010. A New Palladium Precatalyst Allows for the Fast Suzuki−Miyaura Coupling Reactions of Unstable Polyfluorophenyl and 2-Heteroaryl Boronic Acids. J. Am. Chem. Soc.. 132(40):14073-14075. https://doi.org/10.1021/ja1073799
3.
Bruno NC, Tudge MT, Buchwald SL. Design and preparation of new palladium precatalysts for C–C and C–N cross-coupling reactions. Chem. Sci.. 4(3):916-920. https://doi.org/10.1039/c2sc20903a
4.
King RP, Krska SW, Buchwald SL. 2021. A Ligand Exchange Process for the Diversification of Palladium Oxidative Addition Complexes. Org. Lett.. 23(15):6030-6034. https://doi.org/10.1021/acs.orglett.1c02101
5.
McCann SD, Reichert EC, Arrechea PL, Buchwald SL. 2020. Development of an Aryl Amination Catalyst with Broad Scope Guided by Consideration of Catalyst Stability. J. Am. Chem. Soc.. 142(35):15027-15037. https://doi.org/10.1021/jacs.0c06139
6.
Drance MJ, Wang S, Gembicky M, Rheingold AL, Figueroa JS. 2020. Probing for Four-Coordinate Zerovalent Iron in a π-Acidic Ligand Field: A Functional Source of FeL4 Enabled by Labile Dinitrogen Binding. Organometallics. 39(18):3394-3402. https://doi.org/10.1021/acs.organomet.0c00487