Skip to Content
MilliporeSigma

Skip To

760900

Poly(ethylene glycol), 4 acetylene dendron, generation 1

average Mn 6,300

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.23
UNSPSC Code:
12162002


description

acetylene surface groups

form

solid

mol wt

PEG average Mn 6,000 (n~130; esterified with 4 acetylene groups), average Mn 6,300

feature

generation 1

no. Surface Groups

4

functional group

alkyne

General description

Dendrimers and dendrons are symmetrically branched polymer structures that possess a well-defined spatial distribution of functional groups. Dendrimers are chemically modified by attaching poly ethylene glycol (PEG) to increase its solubility and prolong circulation half-life. “Clickable groups”, such as primary acetylenes, can be introduced to the mono dispersed dendritic structures, using click chemistry.

Application

Used as an efficient drug delivery system.


Still not finding the right product?


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters